Learn More
Bioworld NLN Medium, 25 L
Catalog No. 50254758
Encompass_Preferred
NLN MediumContains Micronutrients and Macronutrients as described by Lichter (1982) does not contain Ca(NO3)2.4H2O Add 347 mg/L Ca(NO3)2.4H2O to the already dissolved mediumSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium.Plant tissue culture testedWorking concentration: 1.32g/L (w/o Calcium nitrate); 1.67g/L (w/ Calcium nitrate)Soluble in water
Catalog No. 50-254-758 Supplier Bioworld Supplier No. 306301022
NLN MediumContains Micronutrients and Macronutrients as described by Lichter (1982) does not contain Ca(NO3)2.4H2O Add 347 mg/L Ca(NO3)2.4H2O to the already dissolved mediumSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium.Plant tissue culture testedWorking concentration: 1.32g/L (w/o Calcium nitrate); 1.67g/L (w/ Calcium nitrate)Soluble in water