Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NlpD E.coli His-SU 100ug
SDP

Catalog No. 5000196688
Encompass_Preferred

Recombinant NlpD protein from Escherichia coli, expressed in E. coli and supplied with N-terminal His and SUMO tags. Supplied as a lyophilized powder formulated in PBS (pH 7.4), the product is >95% pure by reducing SDS-PAGE and has an approximate molecular weight of 65 kDa. Intended for biochemical studies of cell wall hydrolase activation, including AmiC-related assays.

  • Recombinant E. coli expression with N-terminal His and SUMO tag.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4.
  • Approximate molecular weight 65 kDa (SDS-PAGE).
  • Recommended reconstitution concentration of at least 100 μg/mL in ddH2O.
  • Store at -20°C; freeze aliquots at -80°C for extended storage.
  • Pack size: 100 μg.

Catalog No. 50-001-96688 Supplier Medchemexpress LLC Supplier No. HYP71498100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant NlpD protein from Escherichia coli, expressed in E. coli and supplied with N-terminal His and SUMO tags. Supplied as a lyophilized powder formulated in PBS (pH 7.4), the product is >95% pure by reducing SDS-PAGE and has an approximate molecular weight of 65 kDa. Intended for biochemical studies of cell wall hydrolase activation, including AmiC-related assays.

  • Recombinant E. coli expression with N-terminal His and SUMO tag.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4.
  • Approximate molecular weight 65 kDa (SDS-PAGE).
  • Recommended reconstitution concentration of at least 100 μg/mL in ddH2O.
  • Store at -20°C; freeze aliquots at -80°C for extended storage.
  • Pack size: 100 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.