Learn More
AdipoGen NLRC5 ms pAb IN113
SDP
Supplier:  AdipoGen AG25B0038C100
Encompass_Preferred
Polyclonal Antibody. Recognizes full-length mouse NLRC5. Does not cross-react with human NLRC5. Application WB. Source Rabbit. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Major histocompatibility complex class I molecules are expressed in all nucleated cells, whereas MHC class II molecules are mostly expressed in hematopoietic cells. The tight regulation of both MHC class I and class II molecules is crucial for eliciting an appropriate immune response. The MHC class II and class I molecule regulation is controlled by class II MHC transactivator CIITA and by NLR family caspase recruitment domain‐containing 5 NLRC5, respectively. NOD-like receptor family CARD domain containing 5 NLRC5 is the largest member of the NOD-like receptors NLRs family.
Catalog No. 50-256-3184