Learn More
AdipoGen NLRP1 NALP1 h pAb AL176
SDP
Supplier:  AdipoGen AG25B0005C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human NLRP1/NALP1. Does not cross-react with mouse NLRP1/NALP1. Source Rabbit. Applications WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. NALP1/NLRP1 is able to form cytoplasmic structures termed death effector filaments. It enhances and stimulates apoptosis through activation of different caspase cascades. It is involved in the activation of caspase-1 and caspase-5 as part of the NALP1 inflammasome complex which leads to processing and release of IL1B and IL18. Genetic variations in the NLRP1/NALP1 gene are associated with susceptibility to vitiligo-associated multiple autoimmune disease type 1, an autoimmune disorder characterized by the association of vitiligo with several autoimmune and autoinflammatory diseases including autoimmune thyroid disease, rheumatoid arthritis and systemic lupus erythematosus.
Catalog No. 50-256-3201