Learn More
Aviva Systems Biology Corp NLRP2 Antibody - C-terminal region (ARP57052_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57052_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NLRP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human NLRP2. Alternate names: CLR19.9, FLJ20510, NALP2, NBS1, PAN1, PYPAF2ApplicationWB. Species reactivity: Human
Catalog No. 50-286-259