Learn More
Selleck Chemical LLC NLRP3/AIM2-IN-3 E5801-25MG
SDP
Catalog No. 5000042713
Encompass_Preferred
NLRP3/AIM2-IN-3 (compound 59) is a potent inhibitor with species-specific effects on NLRP3 and AIM2 inflammasome-mediated pyroptosis It demonstrates potent inhibition of pyroptosis in THP-1 macrophages stimulated with LPS/nigericin with an IC50 of 0 077 M It also disrupts the interaction between NLRP3 or AIM2 and the adaptor protein ASC effectively preventing ASC oligomerization and blocking inflammasome assembly
Catalog No. 50-000-42713 Supplier Selleck Chemical LLC Supplier No. E580125MG
NLRP3/AIM2-IN-3 (compound 59) is a potent inhibitor with species-specific effects on NLRP3 and AIM2 inflammasome-mediated pyroptosis It demonstrates potent inhibition of pyroptosis in THP-1 macrophages stimulated with LPS/nigericin with an IC50 of 0 077 M It also disrupts the interaction between NLRP3 or AIM2 and the adaptor protein ASC effectively preventing ASC oligomerization and blocking inflammasome assembly