Learn More
AdipoGen NLRP3 Antibodies Starter Set
SDP
Supplier:  AdipoGen AG44B0008KI01
Encompass_Preferred
The NLRP3 Inflammasome Human Antibodies Starter Set is an all-in-one solution to study the NLRP3 inflammasome using Western blotting application. This antibody starter set comprises our KO extract validated STANDARD antibodies against the key components of the NLRP3 inflammasome including NLRP3, Asc and Caspase-1, used and published by the experts in inflammasome research. This economic starter set contains enough primary antibodies to perform at least 3 western blot experiments.
Catalog No. 50-256-2750