Learn More
TARGETMOL CHEMICALS INC NLRP3-IN-10 5MG
SDP
Catalog No. 502579522
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NLRP3-IN-10 (ZVN26391) is a potent NLRP3 inhibitor. NLRP3-IN-10 inhibits IL-1(beta) release with an IC50 value of 251.1 nM. NLRP3-IN-10 attenuates ASC speck formation leading to suppress activation of NLRP3 inflammasome. purity: 99%
Catalog No. 50-257-9522 Supplier TARGETMOL CHEMICALS INC Supplier No. T613925MG
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NLRP3-IN-10 (ZVN26391) is a potent NLRP3 inhibitor. NLRP3-IN-10 inhibits IL-1(beta) release with an IC50 value of 251.1 nM. NLRP3-IN-10 attenuates ASC speck formation leading to suppress activation of NLRP3 inflammasome. purity: 99%