Learn More
Medchemexpress LLC HY-W011082 50mg Medchemexpress, NLRP3-IN-2 CAS:16673-34-0 Purity:>98%
SDP
Catalog No. 502083307
Encompass_Preferred
Medchemexpress, HY-W011082 50mg NLRP3-IN-2 CAS:16673-34-0 NLRP3-IN-2, an intermediate substrate in the synthesis of glyburide, inhibits the formation of the NLRP3 inflammasome in cardiomyocytes and limits the infarct size following myocardial ischemia/reperfusion in the mouse, without affecting glucose metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3307 Supplier Medchemexpress LLC Supplier No. HYW01108250MG
Medchemexpress, HY-W011082 50mg NLRP3-IN-2 CAS:16673-34-0 NLRP3-IN-2, an intermediate substrate in the synthesis of glyburide, inhibits the formation of the NLRP3 inflammasome in cardiomyocytes and limits the infarct size following myocardial ischemia/reperfusion in the mouse, without affecting glucose metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.