Learn More
Medchemexpress LLC NLRP3-IN-21 | 2956791-61-8 | 99.0% | 529.32 | 1 ML
SDP
Catalog No. 5000360324
Encompass_Preferred
NLRP3-IN-21 (compound L38) is an NLRP3 inflammasome inhibitor with inflammatory properties. It inhibits NLRP3 inflammasome activation and pyroptosis by suppressing gasdermin D cleavage, ASC oligomerization, and NLRP3 inflammasome assembly.
  • Inhibits NLRP3 inflammasome activation.
  • Suppresses pyroptosis.
  • Reduces gasdermin D cleavage.
  • Prevents ASC oligomerization.
  • Inhibits NLRP3 inflammasome assembly.
Catalog No. 50-003-60324 Supplier Medchemexpress LLC Supplier No. HY14960410MM/1ML
NLRP3-IN-21 (compound L38) is an NLRP3 inflammasome inhibitor with inflammatory properties. It inhibits NLRP3 inflammasome activation and pyroptosis by suppressing gasdermin D cleavage, ASC oligomerization, and NLRP3 inflammasome assembly.
  • Inhibits NLRP3 inflammasome activation.
  • Suppresses pyroptosis.
  • Reduces gasdermin D cleavage.
  • Prevents ASC oligomerization.
  • Inhibits NLRP3 inflammasome assembly.