Learn More
Medchemexpress LLC NLRP3 Mouse His-SU 2ug
SDP
Catalog No. 5000195382
Encompass_Preferred
Recombinant mouse NLRP3 supplied as an N-terminal SUMO fusion with a 6xHis tag, expressed in E. coli and delivered lyophilized. Intended for research use in biochemical and immunological assays, the protein is purified (>95% by reducing SDS-PAGE), has an approximate molecular weight of 35 kDa, and corresponds to residues M1-R153. Storage and reconstitution guidance is provided to maximize stability.
  • Expressed in E. coli as an N-terminal SUMO and 6xHis fusion.
  • Purified to greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight of 35 kDa for assay verification.
  • Lyophilized formulation for convenient storage and transport.
  • Includes recommended storage and reconstitution conditions to preserve stability.
  • Multiple small sizes available to support assay development and pilot studies.
Catalog No. 50-001-95382 Supplier Medchemexpress LLC Supplier No. HYP715982UG
Recombinant mouse NLRP3 supplied as an N-terminal SUMO fusion with a 6xHis tag, expressed in E. coli and delivered lyophilized. Intended for research use in biochemical and immunological assays, the protein is purified (>95% by reducing SDS-PAGE), has an approximate molecular weight of 35 kDa, and corresponds to residues M1-R153. Storage and reconstitution guidance is provided to maximize stability.
  • Expressed in E. coli as an N-terminal SUMO and 6xHis fusion.
  • Purified to greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight of 35 kDa for assay verification.
  • Lyophilized formulation for convenient storage and transport.
  • Includes recommended storage and reconstitution conditions to preserve stability.
  • Multiple small sizes available to support assay development and pilot studies.