Learn More
Aviva Systems Biology Corp NLRP5 Antibody - N-terminal region (ARP59021_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59021_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NLRP5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human NLRP5. Alternate names: CLR19.8, MATER, NALP5, PAN11, PYPAF8ApplicationWB. Species reactivity: Human
Catalog No. 50-286-562