Learn More
AdipoGen anti-NLRP6/NALP6 (human), mAb (Clint-1) (100 µg)
SDP
Supplier:  AdipoGen AG20B0046C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human NLRP6. Isotype: Mouse IgG1kappa. Clone: Clint-1. Applications: IHC, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. The inflammasome is a multiprotein complex that mediates the activation of caspase-1, which promotes, amongst others, the secretion of the proinflammatory cytokines interleukin (IL)-1beta and IL-18. Members of the Nod-like receptor (NLR) family are critical components of the inflammasome that link danger-signals to caspase-1 activation. NLRP6/NALP6 is important for intestinal homeostasis by sensing pathogenic commensal bacteria and activating bactericidal activity of macrophages through secretion of IL-18. NLRP6 suppresses inflammation and carcinogenesis by regulating tissue repair and deletion of NLRP6 gene accelerated colitis-associated tumor growth in mice.
Catalog No. 50-205-2957