Learn More
Medchemexpress LLC Nls (pkkkrkv) | 95088-49-6 | 99.0% | 883.14 | 5 MG
SDP
Catalog No. 5000423187
Encompass_Preferred
NLS (PKKKRKV) is a nuclear localization signal (NLS) derived from the SV40 large T antigen, mediating the binding of karyophilic proteins to importin α. It enhances nuclear entry in gene transfer research.
  • Enhances nuclear entry in gene transfer research
  • Reduces transfection efficiency in 293T cells with pmaxGFP plasmid pre-incubation
  • Significantly enhances transfection efficiency in 293T cells with DSPC pre-incubation
Catalog No. 50-004-23187 Supplier Medchemexpress LLC Supplier No. HYP18765MG
NLS (PKKKRKV) is a nuclear localization signal (NLS) derived from the SV40 large T antigen, mediating the binding of karyophilic proteins to importin α. It enhances nuclear entry in gene transfer research.
  • Enhances nuclear entry in gene transfer research
  • Reduces transfection efficiency in 293T cells with pmaxGFP plasmid pre-incubation
  • Significantly enhances transfection efficiency in 293T cells with DSPC pre-incubation