Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nls (pkkkrkv) (hydrochloride) | 99.0% | 919.60 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYP1876B

Encompass_Preferred

NLS (PKKKRKV) hydrochloride is the hydrochloride form of NLS (PKKKRKV). It is a nuclear localization signal (NLS) derived from the simian virus 40 large tumor antigen (SV40 large T antigen). This signal mediates the binding of karyophilic protein to importin α and can enhance nuclear entry in gene transfer research.

  • Derived from simian virus 40 large tumor antigen (SV40 large T antigen).
  • Mediates binding of the karyophilic protein to importin α.
  • Enhances nuclear entry in gene transfer research.
  • Appearance: Solid.
  • Color: White to off-white.
  • Sequence: Pro-Lys-Lys-Lys-Arg-Lys-Val.

Catalog No. 50-004-26834


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.