Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nls (pkkkrkv) (hydrochloride) | 99.0% | 919.60 | 50 MG
SDP

Catalog No. 5000424308
Encompass_Preferred

NLS (PKKKRKV) hydrochloride is the hydrochloride form of NLS (PKKKRKV). It is a nuclear localization signal (NLS) derived from the simian virus 40 large tumor antigen (SV40 large T antigen). This NLS mediates the binding of karyophilic protein to importin α and can enhance nuclear entry in gene transfer research.

  • Solid appearance.
  • White to off-white color.
  • Sequence: Pro-Lys-Lys-Lys-Arg-Lys-Val.
  • Enhances nuclear entry in gene transfer research.
  • Mediates binding of karyophilic protein to importin α.
  • Recommended storage: powder at -80°c for 2 years or -20°c for 1 year; in solvent at -80°c for 6 months or -20°c for 1 month. Keep sealed, away from moisture and light.

Catalog No. 50-004-24308 Supplier Medchemexpress LLC Supplier No. HYP1876B50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NLS (PKKKRKV) hydrochloride is the hydrochloride form of NLS (PKKKRKV). It is a nuclear localization signal (NLS) derived from the simian virus 40 large tumor antigen (SV40 large T antigen). This NLS mediates the binding of karyophilic protein to importin α and can enhance nuclear entry in gene transfer research.

  • Solid appearance.
  • White to off-white color.
  • Sequence: Pro-Lys-Lys-Lys-Arg-Lys-Val.
  • Enhances nuclear entry in gene transfer research.
  • Mediates binding of karyophilic protein to importin α.
  • Recommended storage: powder at -80°c for 2 years or -20°c for 1 year; in solvent at -80°c for 6 months or -20°c for 1 month. Keep sealed, away from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.