Learn More
Cayman Chemical nlumbo nucfera Dphyln 10mg
SDP
Catalog No. 502940398
Encompass_Preferred
A lignan with diverse biological activities; reduces viral titers in SARS-CoV-2-infected Vero cells (EC50 = 1.92 µM); inhibits bovine chromaffin granule V-ATPase (IC50 = 17 nM) and lysosomal acidification in primary human osteoclasts; induces apoptosis in and inhibits the growth of SGC-7901 cells at 1, 5, and 10 µM; reduces intratumoral expression of V-ATPase and tumor volume in an SGC-7901 mouse xenograft model at 20 mg/kg
Catalog No. 50-294-0398 Supplier Cayman Chemical Supplier No. 2782610 MG
A lignan with diverse biological activities; reduces viral titers in SARS-CoV-2-infected Vero cells (EC50 = 1.92 µM); inhibits bovine chromaffin granule V-ATPase (IC50 = 17 nM) and lysosomal acidification in primary human osteoclasts; induces apoptosis in and inhibits the growth of SGC-7901 cells at 1, 5, and 10 µM; reduces intratumoral expression of V-ATPase and tumor volume in an SGC-7901 mouse xenograft model at 20 mg/kg