Learn More
Cayman Chemical Nuciferine, Purity: ≥98%, CAS: 475-83-2, 5 mg
SDP
Catalog No. 502946362
Encompass_Preferred
An alkaloid with diverse biological activities; partial agonist at dopamine D2 receptors, as well as an inverse agonist at the serotonin receptor subtype 5-HT7, and an antagonist at the 5-HT2B and 5-HT2C receptors (EC50s = 65.07, 150, 478, and 131 nM, respectively); an antagonist at schistosome Sm.5-HTRL receptors (IC50 = 240 nM); inhibits 5-HT-induced motility of larval and adult S. mansoni at 10 µM; activates PPARα, PPARβ/δ, and PPARγ transcriptional activity; prevents RSL3-induced ferroptotic cell death in HK-2 and HEK293T cells at 10, 20, and 40 µM; reduces serum hyperlipidemia and hepatic steatosis in a mouse model of high-fat diet-induced hyperlipidemia
Catalog No. 50-294-6362 Supplier Cayman Chemical Supplier No. 338395 MG
An alkaloid with diverse biological activities; partial agonist at dopamine D2 receptors, as well as an inverse agonist at the serotonin receptor subtype 5-HT7, and an antagonist at the 5-HT2B and 5-HT2C receptors (EC50s = 65.07, 150, 478, and 131 nM, respectively); an antagonist at schistosome Sm.5-HTRL receptors (IC50 = 240 nM); inhibits 5-HT-induced motility of larval and adult S. mansoni at 10 µM; activates PPARα, PPARβ/δ, and PPARγ transcriptional activity; prevents RSL3-induced ferroptotic cell death in HK-2 and HEK293T cells at 10, 20, and 40 µM; reduces serum hyperlipidemia and hepatic steatosis in a mouse model of high-fat diet-induced hyperlipidemia