Learn More
Cayman Chemical Lotusine, Purity: ≥98%, CAS: 6871-67-6, 50 mg
SDP
Catalog No. 502911472
Encompass_Preferred
An alkaloid with diverse biological activities; increases cAMP levels in rat myocardium at 0.1-3 µM and cGMP levels in rabbit corpus cavernosum in a concentration-dependent manner; prevents doxorubicin-induced lipid peroxidation in H9c2 embryonic rat cardiomyocytes; increases LVP, LVEDP, and SAP in anesthetized rats at 5 mg/kg
Catalog No. 50-291-1472 Supplier Cayman Chemical Supplier No. 3513650 MG
An alkaloid with diverse biological activities; increases cAMP levels in rat myocardium at 0.1-3 µM and cGMP levels in rabbit corpus cavernosum in a concentration-dependent manner; prevents doxorubicin-induced lipid peroxidation in H9c2 embryonic rat cardiomyocytes; increases LVP, LVEDP, and SAP in anesthetized rats at 5 mg/kg