Learn More
Selleck Chemical LLC NLX-101 S3510-5mg
SDP
Catalog No. 503110583
Encompass_Preferred
NLX-101 (F-15599) is a highly selective 5-HT1A receptor biased agonist that mediates antidepressant-like activity in rats via prefrontal cortex 5-HT 1A receptors
Catalog No. 50-311-0583 Supplier Selleck Chemical LLC Supplier No. S35105MG
NLX-101 (F-15599) is a highly selective 5-HT1A receptor biased agonist that mediates antidepressant-like activity in rats via prefrontal cortex 5-HT 1A receptors