Learn More
Waters Corp Waters Ferrule, PEEK, 1/16 in OD, HPLC
Catalog No. 50818737 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ferrule is engineered to create leak-free connections, ensuring low dead volume and minimal band spreading within your system. Optimal performance requires that these components are clean and harmonized. The ferrule’s geometry must precisely match the conical port it’s used with, and the tube’s diameter must correspond to the fittings, offering flexibility for system setup and maintenance.
  • Leak-free connections
  • Low dead volume
  • Minimal band spreading
Catalog No. 50-818-737 Supplier Waters Corp Supplier No. WAT021817
This ferrule is engineered to create leak-free connections, ensuring low dead volume and minimal band spreading within your system. Optimal performance requires that these components are clean and harmonized. The ferrule’s geometry must precisely match the conical port it’s used with, and the tube’s diameter must correspond to the fittings, offering flexibility for system setup and maintenance.
  • Leak-free connections
  • Low dead volume
  • Minimal band spreading