Learn More
Medchemexpress LLC HY-10468 5mg Medchemexpress, NM107 CAS:20724-73-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY104685MG
Encompass_Preferred
Medchemexpress, HY-10468 5mg NM107 CAS:20724-73-6 NM107 (2'-C-Methylcytidine) is an nucleoside inhibitor of the hepatitis C virus (HCV) NS5B polymerase, the EC50 of NM107 in the wild-type replicon cells is 1.85 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8229