Learn More
Eurofins Discoverx PathHunter NMBR Total Internalization U2OS Cell Line, 2 vials. (93-0939C3)
Supplier:  Eurofins Discoverx 930939C3
Encompass
The PathHunter NMBR Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of NMBR via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged NMBR and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the NMBR receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1614
Add to Cart
Request a Quote