Learn More
Aviva Systems Biology Corp NMD3 Antibody - C-terminal region (ARP64485_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64485_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NMD3. It was validated on Western Blot. . Alternate names: CGI-07, FLJ21053ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-274