Learn More
TARGETMOL CHEMICALS INC NMDA 200MG
SDP
Catalog No. 502576485
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. N-Methyl-D-aspartic acid is an amino acid that as the D-isomer is the defining agonist for the NMDA (N-Methyl-D-aspartic acid) receptor subtype of glutamate receptors. purity: 99%
Catalog No. 50-257-6485 Supplier TARGETMOL CHEMICALS INC Supplier No. T6608200MG
Also available in 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. N-Methyl-D-aspartic acid is an amino acid that as the D-isomer is the defining agonist for the NMDA (N-Methyl-D-aspartic acid) receptor subtype of glutamate receptors. purity: 99%