Learn More
Medchemexpress LLC HY-17551 50mg , NMDA CAS:6384-92-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1755150MG
Encompass_Preferred
HY-17551 50mg NMDA CAS: 6384-92-5 NMDA is a specific agonist for NMDA receptor mimicking the action of glutamate, the neurotransmitter which normally acts at that receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9380