Learn More
Medchemexpress LLC HY-17551 100mg Medchemexpress, NMDA N-Methyl-D-aspartic acid CAS:6384-92-5 Purity:98%
SDP
Catalog No. 501638674
Encompass_Preferred
Medchemexpress,HY-17551 100mg NMDA N-Methyl-D-aspartic acid CAS:6384-92-5 Formula:C5H9NO4 NMDA is a specific agonist for NMDA receptor mimicking the action of glutamate, the neurotransmitter which normally acts at that receptor. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8674 Supplier Medchemexpress LLC Supplier No. HY17551100MG
Medchemexpress,HY-17551 100mg NMDA N-Methyl-D-aspartic acid CAS:6384-92-5 Formula:C5H9NO4 NMDA is a specific agonist for NMDA receptor mimicking the action of glutamate, the neurotransmitter which normally acts at that receptor. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.