Learn More
Cayman Chemical NMDA Receptor NR2B Subunit (Phospho-Tyr1252) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502886536
Encompass_Preferred
Antigen: phosphopeptide corresponding to amino acid residues surrounding phospho-Tyr1252 of the human NMDA Receptor NR2B Subunit • Host: rabbit • Cross Reactivity: (+) human and rat NMDA receptor; expected to react with bovine, canine, chicken, mouse, non-human primates, and zebrafish NMDA receptor • Application(s): WB • Phosphorylation of Tyr1252 in NR2B is thought to potentiate NMDA receptor-dependent influx of calcium.
Catalog No. 50-288-6536 Supplier Cayman Chemical Supplier No. 100097591 EA
Antigen: phosphopeptide corresponding to amino acid residues surrounding phospho-Tyr1252 of the human NMDA Receptor NR2B Subunit • Host: rabbit • Cross Reactivity: (+) human and rat NMDA receptor; expected to react with bovine, canine, chicken, mouse, non-human primates, and zebrafish NMDA receptor • Application(s): WB • Phosphorylation of Tyr1252 in NR2B is thought to potentiate NMDA receptor-dependent influx of calcium.