Learn More
Cayman Chemical NMDA Receptor NR2B Subunit (Phospho-Tyr1472) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502886856
Encompass_Preferred
Antigen: phosphopeptide corresponding to amino acid residues surrounding phospho-Tyr1472 of NMDA Receptor NR2B Subunit • Host: rabbit • Cross Reactivity: (+) human and rat NMDA receptor; expected to react with bovine, canine, chicken, mouse, non-human primates, and zebrafish NMDA receptor • Application(s): WB • Phosphorylation of Tyr1472 on NR2B may regulate the functional expression of the receptor in LTP and other forms of plasticity.
Catalog No. 50-288-6856 Supplier Cayman Chemical Supplier No. 100097611 EA
Antigen: phosphopeptide corresponding to amino acid residues surrounding phospho-Tyr1472 of NMDA Receptor NR2B Subunit • Host: rabbit • Cross Reactivity: (+) human and rat NMDA receptor; expected to react with bovine, canine, chicken, mouse, non-human primates, and zebrafish NMDA receptor • Application(s): WB • Phosphorylation of Tyr1472 on NR2B may regulate the functional expression of the receptor in LTP and other forms of plasticity.