Learn More
Cayman Chemical NMDA Receptor NR2B Subunit Polyclonal Antibody, 1 ea
SDP
Catalog No. 502886832
Encompass_Preferred
Antigen: fusion protein from the C-terminus of the NR2B subunit of rat NMDA receptor • Host: rabbit • Cross Reactivity: (+) human, rat, and mouse NMDA receptor • Application(s): IHC, IP, and WB • Overexpression of the NR2B-subunit of the NMDA receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression.
Catalog No. 50-288-6832 Supplier Cayman Chemical Supplier No. 106091 EA
Antigen: fusion protein from the C-terminus of the NR2B subunit of rat NMDA receptor • Host: rabbit • Cross Reactivity: (+) human, rat, and mouse NMDA receptor • Application(s): IHC, IP, and WB • Overexpression of the NR2B-subunit of the NMDA receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression.