Learn More
Medchemexpress LLC HY-111500 10mg Medchemexpress, NMDAR antagonist 1 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11150010MG
Encompass_Preferred
Medchemexpress, HY-111500 10mg NMDAR antagonist 1 CAS: NMDAR antagonist 1 is a potent and orally bioavailable NR2B-selective NMDAR antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8890