Learn More
Aviva Systems Biology Corp Nme1 Antibody - C-terminal region (ARP38169_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38169_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nme1. It was validated on Western Blot. . Alternate names: AL024257, NDPK-A, NM23-M1, NM23AApplicationDB, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-284-726