Learn More
ACROBiosystems Recombinant Protein;Human NME1 / NDKA / NM23-H1 Protein;E.coli;1MG;NM1,NME1,NDK A,NDP kinase A,NDPA,GAAD,NM23-H1
Supplier:  ACROBiosystems NM1H51471MG
Encompass_Preferred
Recombinant Protein;Human NME1 / NDKA / NM23-H1 Protein;1MG;Human NME1, His Tag (NM1-H5147) is expressed from E.coli cells. It contains AA Ala 2 - Glu 152 (Accession # AAH00293).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18 kDa. The protein migrates as 21 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8882