Learn More
New England Biolabs, Inc. NmeAIII – 250 units
SDP
Catalog No. 50995289
Encompass
A restriction endonuclease that recognizes the sequence GCCGAGNNNNNNNNNNNNNNNNNNN_NN^.
Catalog No. 50-995-289 Supplier New England Biolabs, Inc. Supplier No. R0711S
A restriction endonuclease that recognizes the sequence GCCGAGNNNNNNNNNNNNNNNNNNN_NN^.