Learn More
Cayman Chemical N-methyl-2-Phenylpropan-1-amine (hydrochloride), Purity: ≥98%, CAS: 5969-39-1, 50 mg
SDP
Catalog No. 502968039
Encompass_Preferred
A phenethylamine that is a positional isomer of methamphetamine
Catalog No. 50-296-8039 Supplier Cayman Chemical Supplier No. 1856350 MG
A phenethylamine that is a positional isomer of methamphetamine