Learn More
Cayman Chemical N-Methyl-4-pyridone-5-carboxamide, Purity: ≥95%, CAS: 769-49-3, 50 mg
SDP
Catalog No. 502910011
Encompass_Preferred
A metabolite of nicotinic acid and a uremic toxin; plasma levels are increased and positively correlated with serum levels of MDA in patients with CKD
Catalog No. 50-291-0011 Supplier Cayman Chemical Supplier No. 3668450 MG
A metabolite of nicotinic acid and a uremic toxin; plasma levels are increased and positively correlated with serum levels of MDA in patients with CKD