Learn More
Cayman Chemical NmethylProtoporphyrInIX500ug
SDP
Catalog No. 502982030
Encompass_Preferred
A transition state analog that potently inhibits protoporphyrin IX ferrochelatase (Ki = 10 nM), blocking the terminal step of the heme biosynthetic pathway
Catalog No. 50-298-2030 Supplier Cayman Chemical Supplier No. 20846500 UG
A transition state analog that potently inhibits protoporphyrin IX ferrochelatase (Ki = 10 nM), blocking the terminal step of the heme biosynthetic pathway