Learn More
Perkin Elmer US LLC NM-H2 Plus Hydrogen Generator - 100 mL/min
Catalog No. 502112956
Encompass_Preferred
Ideal for operation with gas analyzers, as fuel gas for flame tools, or as a source for pure hydrogen in plasma chambers and other isolated environments.
Catalog No. 50-211-2956 Supplier Perkin Elmer US LLC Supplier No. N9308581
Add to Cart
Add to Cart
Request a Quote
Ideal for operation with gas analyzers, as fuel gas for flame tools, or as a source for pure hydrogen in plasma chambers and other isolated environments.