Learn More
Aviva Systems Biology Corp NMI Antibody - N-terminal region (ARP35736_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35736_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NMI. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NMI. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55833