Learn More
Aviva Systems Biology Corp NMI Antibody - middle region (ARP35737_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35737_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NMI. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NMI. Alternate names: -ApplicationIHC, WB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55834