Learn More
Enzo Life Sciences NMNAT3 (human), (recombinant) (His-tag) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX201238C100
Encompass_Preferred
Alternative name: Nicotinamide mononucleotide adenylyltransferase 3, NMN adenylyltransferase 3. Purity: ≥95% (SDS-PAGE). MW: 29kDa (monomer). Formulation: Liquid. In 50mM TRIS-HCl, pH 8.0, containing 300mM sodium chloride, 1mM DTT and 10% glycerol. Source: Produced in E. coli. Full length human NMNAT3 is fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7953