Learn More
Wilmad Labglass NMR TBE ECON 5MMX7 400MHZ 5PK
Catalog No. 50802498
Encompass_Preferred
NMR Tube, Economy Brand, Thin Walled, 5mm O.D. x 7 in. length. MHz Rating: 400; Wall Thickness: 0.38mm; Concentricity: 0.0010 in. (average); Camber: 0.0005 in. (average). Manufactured from 51-expansion borosilicate glass. 5/pack.
Catalog No. 50-802-498 Supplier Wilmad Labglass Supplier No. WG122875
NMR Tube, Economy Brand, Thin Walled, 5mm O.D. x 7 in. length. MHz Rating: 400; Wall Thickness: 0.38mm; Concentricity: 0.0010 in. (average); Camber: 0.0005 in. (average). Manufactured from 51-expansion borosilicate glass. 5/pack.