Learn More
Wilmad Labglass 5 mm NMR Sample Tube, Type 1 Class B Glass, 7" L 100/Pack, High-Throughput
Catalog No. NC0921373
Encompass_Preferred
5 mm NMR Economy Sample Tube w/ Cap, 7 in. L 100 Pk, High-Throughput. Wilmad-LabGlass High-Throughput NMR Tubes are manufactured from borosilicate glass that meets the aSTM Type 1 Class B standard and recognized as N51a. Due to its nature, this glass has much less thermal-shock resistance than Class a glass used for our Precision Tubes. It may contain Fe2O3 which is paramagnetic around 1200 ppm and has certain impact on shimming quality at high field over 600 MHz. High-Throughput NMR tubes have an averaged camber of 60 microns to guarantee the spectrum resolution for small molecule (MW<250) samples up to 600 MHz. WG-1000 Tubes are sold in various pack sizes and include the corresponding quantity of Blue Caps. Other colors are available for purchase separately. The twist pack is made of polypropylene and resistant to common organic solvents. The packing materials are 100% recyclable.
Catalog No. NC0921373 Supplier Wilmad Labglass Supplier No. WG10007
5 mm NMR Economy Sample Tube w/ Cap, 7 in. L 100 Pk, High-Throughput. Wilmad-LabGlass High-Throughput NMR Tubes are manufactured from borosilicate glass that meets the aSTM Type 1 Class B standard and recognized as N51a. Due to its nature, this glass has much less thermal-shock resistance than Class a glass used for our Precision Tubes. It may contain Fe2O3 which is paramagnetic around 1200 ppm and has certain impact on shimming quality at high field over 600 MHz. High-Throughput NMR tubes have an averaged camber of 60 microns to guarantee the spectrum resolution for small molecule (MW<250) samples up to 600 MHz. WG-1000 Tubes are sold in various pack sizes and include the corresponding quantity of Blue Caps. Other colors are available for purchase separately. The twist pack is made of polypropylene and resistant to common organic solvents. The packing materials are 100% recyclable.