Learn More
Wilmad Labglass NMR TBE PREC 5MMX7 500MHZ 5PK
Catalog No. 50802531
Encompass_Preferred
Tube, NMR; Wilmad Labglass; Precision; Thin wall; 5mm O.D. x 7 in. L; MHz Rating: 500; Wall thickness: 0.38mm; Concentricity: 0.001 in. (T.I.R.); Camber: 0.0005 in. (T.I.R.); Pyrex or equivalent glass; 5/pack.
Catalog No. 50-802-531 Supplier Wilmad Labglass Supplier No. 528PP75
Tube, NMR; Wilmad Labglass; Precision; Thin wall; 5mm O.D. x 7 in. L; MHz Rating: 500; Wall thickness: 0.38mm; Concentricity: 0.001 in. (T.I.R.); Camber: 0.0005 in. (T.I.R.); Pyrex or equivalent glass; 5/pack.