Learn More
Chemglass Life Sciences NMR Tube Cap, 5mm, Red
SDP
Catalog No. CG184202
Encompass_Preferred
NMR Tube Cap, 5mm, Red. Made from polyethylene, these caps are sized to fit 5mm O.D. NMR sample tubes. Cap fits tightly to stay in place during increased internal vaporization pressure. Available in a pack of 100, or in a convenient divided polypropylene box having 100 each of the 6 listed colors.
Catalog No. CG184202 Supplier Chemglass Life Sciences Supplier No. CG184202
NMR Tube Cap, 5mm, Red. Made from polyethylene, these caps are sized to fit 5mm O.D. NMR sample tubes. Cap fits tightly to stay in place during increased internal vaporization pressure. Available in a pack of 100, or in a convenient divided polypropylene box having 100 each of the 6 listed colors.