Learn More
Chemglass Life Sciences NMR Tube Cleaner, Morris, For 5mm X 9in NMR Tube, Complete
SDP
Catalog No. CG185003
Encompass_Preferred
NMR Tube Cleaner, Morris, For 5mm X 9in NMR Tube, Complete. The Morris NMR tube cleaner is an inexpensive, durable apparatus which will clean 5mm NMR tubes with as little as 10mL of solvent in one easy operation. Cleaner does not require a stopper on the NMR tube and does not have a fragile side arm. Complete item is supplied with a NMR tube washer, adapter, 250ML HW filter flask and instructions for use.**Designed and developed by Mr. Earl Morris, Georgetown University, Washington DC
Catalog No. CG185003 Supplier Chemglass Life Sciences Supplier No. CG185003
NMR Tube Cleaner, Morris, For 5mm X 9in NMR Tube, Complete. The Morris NMR tube cleaner is an inexpensive, durable apparatus which will clean 5mm NMR tubes with as little as 10mL of solvent in one easy operation. Cleaner does not require a stopper on the NMR tube and does not have a fragile side arm. Complete item is supplied with a NMR tube washer, adapter, 250ML HW filter flask and instructions for use.**Designed and developed by Mr. Earl Morris, Georgetown University, Washington DC