Learn More
Chemglass Life Sciences Tube, NMR, Valved, For 500 MHz, 7" Long
SDP
Catalog No. NC1014473
Encompass_Preferred
J Young Valve NMR Tubes are is easy to use and to disassemble for cleaning. It includes a female joint for quick attachment to your vacuum rack O-Ring
Catalog No. NC1014473 Supplier Chemglass Life Sciences Supplier No. CS5500JY7
J Young Valve NMR Tubes are is easy to use and to disassemble for cleaning. It includes a female joint for quick attachment to your vacuum rack O-Ring