Learn More
Sigma Aldrich Fine Chemicals Biosciences NMR tube labels temperatur
Catalog No. 502901863
Encompass
Self-laminating vinyl labels with white write-on surface resist oil water and solvent. Label wraps around 5mm tube for positive sample identification. Supplied with 26 labels on a card 25 cards per pack.
Catalog No. 50-290-1863 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2205741PAK
Self-laminating vinyl labels with white write-on surface resist oil water and solvent. Label wraps around 5mm tube for positive sample identification. Supplied with 26 labels on a card 25 cards per pack.