Learn More
Norell Inc NMR TUBES 1.7MM OD 103.5MM
SDP
Catalog No. NC2384259
Encompass_Preferred
1.7mm OD, 103.5mm LONG, RATED UP TO 700MHz, SAMPLE VAULT SERIES NMR TUBES FOR AUTOMATED SAMPLE TUBE SYSTEM, 96PK, INCLUDES OPEN PORT CODED SAMPLE VAULT CAPS, INCLUDES SAMPLE VAULT™ NMR SAMPLE TUBE RACK
Catalog No. NC2384259 Supplier Norell Inc Supplier No. SVOP1.7103.596PK
1.7mm OD, 103.5mm LONG, RATED UP TO 700MHz, SAMPLE VAULT SERIES NMR TUBES FOR AUTOMATED SAMPLE TUBE SYSTEM, 96PK, INCLUDES OPEN PORT CODED SAMPLE VAULT CAPS, INCLUDES SAMPLE VAULT™ NMR SAMPLE TUBE RACK