Learn More
Norell Inc 5mm OD, 103.5mm LONG, RATED UP TO 700MHz, SAMPLE VAULT SERIES NMR TUBES FOR AUTOMATED SAMPLE TUBE SYSTEM, 96PK, INCLUDES CLOSED PORT SAMPLE VAULT CAPS
SDP
Catalog No. NC1843967
Encompass_Preferred
5mm OD, 103.5mm LONG, RATED UP TO 700MHz, SAMPLE VAULT SERIES NMR TUBES FOR AUTOMATED SAMPLE TUBE SYSTEM, 96PK, INCLUDES CLOSED PORT SAMPLE VAULT CAPS
Catalog No. NC1843967 Supplier Norell Inc Supplier No. SVCP5103.596PK
5mm OD, 103.5mm LONG, RATED UP TO 700MHz, SAMPLE VAULT SERIES NMR TUBES FOR AUTOMATED SAMPLE TUBE SYSTEM, 96PK, INCLUDES CLOSED PORT SAMPLE VAULT CAPS